Luer to Hose Barb Fitting
Catalog # COLE50114-69
Supplier: Cole-Parmer
New Product
Specifications
- Connection Type:Male Luer to hose barb
- Description:Luer to hose barb fitting, straight adapter
- Material:PP
- Pk:10
- Barb size (port 2):3,175 mm
Specifications
About this item
Cole-Parmer Luer to hose barb fitting are manufactured in an ISO Class 7 certified cleanroom to minimise contamination by pollutants.
- Two ports
- Male luer ×3,175 mm hose barb
- Animal-derivative-free
- Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
- Free technical application support available to determine which connector type is needed for your application
- PVC-free - ideal for use in bioprocess, life science, and pharmaceutical industries
- Connect to a female luer body to form a leak-free fluid path
- Single barb design tightly 'grips' the tubing
Certifications: FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant, ADCF, non-DEHP; non-phthalate.