
Xylenol orange triturate 1% (w/w) in potassium nitrate Reag. Ph. Eur. 1096301
Catalog # 87963.150
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Test for sensitivity:passes test
Specification Test Results
About this item
:
:672.67 g/mol
Storage Temperature:Ambient
EINECS:216-553-8
UN:1486
ADR:5.1,III
:GHS03
CAS Number:1611-35-4
Formula:C₃₁H₃₂N₂O₁₃S
MDL Number:MFCD00014580