Water, WFI (water for injection) quality, sterile VAI®
Catalog # VELTVAI-WFI-2G
Supplier: VELTEK ASSOCIATES
New Product
CAS Number:
Specifications
- Pack type:Bottle
- Pk:2 Bottle
- Seq:3107
- Environmentally Preferable:
- Pk Info:7,6 L (256 oz.)
Specifications
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
Merck Index:13,10098