X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Catalog # MOLEM30473975
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:408.63 g/mol
Melting Pt:237…239 °C
Storage Temperature:Freezer
EINECS:230-640-8
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666