3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Supplier: Serva
Certificates
About this item
Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
CAS Number:75621-03-3
:
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083
:614.89 g/mol
Merck Index:13,02052
Melting Pt:116 °C
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together