
N,N-Diethyl-p-phenylenediammonium sulphate, Supelco®
Catalog # 07670-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:262.33 g/mol
Boiling Pt:274…275 °C (1013 hPa)
Melting Pt:184…186 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:228-500-6
:GHS07
CAS Number:6283-63-2
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993