- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:C-Myc/DDK
- Storage conditions:Store at −80 °C
- Protein synonyms:PP2C-beta|PPC2BETAX|PP2CBETA|PP2CB|PP2C-beta-X
- UniProtKB:O75688
- Protein/peptide name:PPM1B (NM_177968)
- Purity:>80% as determined by SDS-PAGE and Coomassie blue staining
- Molecular weight:42,6 kDa
- Concentration:>0,05 µg/µl as determined by microplate BCA method
Recombinant protein of human protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform (PPM1B), transcript variant 2, 100 µg.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control.
Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.
Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol
Preparation: Recombinant protein was captured through anti-DDK affinity column followed by conventional chromatography steps
Stability: Stable for 12 months from the date of receipt of the product under proper storage and handling conditions, avoid repeated freeze-thaw cycles
For testing in cell culture applications, please filter before use. Note that you may experience some loss of protein during the filtration process.