Anti-Adenosine A2a R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7F6-G5-A2]
NOVUNBP1-39474PE
New Product
- Antibody type:Primary
- Antigen name:Adenosine A2a R
- Clonality:Monoclonal
- Clone:7F6-G5-A2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Pig,Rabbit,Human,Guinea pig,Dog,Rat,Mouse
- Epitope:Epitope mapped to SQPLPGER in the third intracellular loop (PMID 9822147)
- Gene ID:135
- Antigen synonyms:adenosine A2 receptor|adenosine A2a receptor|adenosine receptor subtype A2a|ADORA2|hA2aR|adenosine receptor A2a|ADORA 2|RDC8
- UniProtKB:P29274
- Storage buffer:PBS
- Storage temperature:Store at 4 °C in the dark
- Immunogen:Full length recombinant human Adenosine A2a R. [UniProt# P29274]
- Purification:Protein G purified
- Pk:0,1 mL
The Adenosine A2a R Antibody (7F6-G5-A2) [PE] from Novus Biologicals is a mouse monoclonal antibody to Adenosine A2a R. This antibody reacts with human, mouse, rat, porcine, canine, guinea pig, rabbit. The Adenosine A2a R Antibody (7F6-G5-A2) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: Adenosine A2a R
Clonality: Monoclonal
Clone: 7F6-G5-A2
Conjugation: PE (Phycoerythrin)
Epitope: Epitope mapped to SQPLPGER in the third intracellular loop (PMID 9822147)
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit