Supelco® Organic Reference Standard, Palmitic Acid-1,2,3,4-¹³C₄
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:287100-89-4
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00190320
- M-Clarity quality level:MQ100
- Synonym(s):Hexadecanoic acid-1,2,3,4-¹³C₄
- Melting point:61...64 °C (lit.)
- Formula:CH₃(CH₂)₁₁(¹³CH₂)₃¹³CO₂H
- Molecular weight:260,39
Specifications
Frequently Bought Together