Supelco® TraceCERT® Organic Reference Standard, L-Ornithine MonohydroChloride
Catalog # 19943-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- CAS No.:3184-13-2
- Description:ʟ-Ornithine monohydrochloride
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00064562
- M-Clarity quality level:MQ300
- Synonym(s):(S)-2,5-Diaminopentanoic acid monohydrochloride
- Molecular weight:168,62
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.