Supelco® Organic Reference Standard, F.A.M.E. Mix C8 - C22
Catalog # CRM18920
Supplier: Merck
CAS Number:
Specifications
- Concentration:Neat
- Pk:1
- Standards:Certified reference material
- Description:F.A.M.E. mix C8 - C22
- Storage conditions:–10...–25 °C
- Hazard class description:H318 - H410
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Synonym(s):Fatty Acid Methyl Ester Mix
- Pk Info:Ampule of 100 mg
Specifications
About this item
- GHS signal: GHS05
Caution: P273 - P280 - P305 + P351 + P338 - P391 - P501