Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Cyclin E2 Rabbit Polyclonal Antibody
  ANTIA10048-100
New Product
undefined
Anti-Cyclin E2 Rabbit Polyclonal Antibody
  ANTIA10048-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    G1/S specific cyclin E2
  • Antigen symbol:
    Cyclin E2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O96020
  • Antigen synonyms:
    CCNE 2|CyclinE2|CYCE2|CYCE 2|CCNE2 protein|CCN E2|CYC E2|Cyclin E2|CCNE2
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    47 kDa
  • Sequence:
    ALKDAPKVLLPQYSQETFIQIAQLLDLCILAIDSLEFQYRILTAAALCHFTSIEVVKKASGLEWDSISECVDWMVPFVNVVKSTSPVKLKTFKKIPMEDRHNIQTHTNYLAMLEEVNYINTFRKGGQLSPVCNGGIMTPPKSTEKPPGKH
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 255 - 404 of human Cyclin E2 (NP_477097.1)
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Cyclin E2 for WB and ICC/IF with samples derived from human.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Cyclin E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human