Anti-Adrenomedullin Rabbit Polyclonal Antibody
Catalog # ANTIA100063-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Adm
- Antigen symbol:Adrenomedullin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P35318
- Antigen synonyms:ADM precursor|Proadrenomedullin N20 terminal peptide|AM|Proadrenomedullin N-20 terminal peptide|ADML_HUMAN|Adrenomedullin|PAMP|ProAM N terminal 20 peptide|Preproadrenomedullin
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human ADM (amino acids 101-150)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ADM for WB and ELISA with samples derived from human and rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Adrenomedullin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat