Anti-ATF2 (phospho Thr69 + Thr71) Rabbit Polyclonal Antibody
ANTIA93029-100
New Product
- Antibody type:Primary
- Antigen name:Activating transcription factor 2
- Antigen symbol:ATF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P15336
- Antigen synonyms:cAMP response element binding protein CRE BP1|cAMP Response Element Binding Protein 2|ATF2 protein|ATF 2|Atf-2|ATF2|cAMP response element-binding protein CRE-BP1|ATF2_HUMAN|Activating transcription factor 2 splice variant ATF2 var2
- Modification:phospho Thr69 + Thr71
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
- Molecular weight:70 kDa
- Sequence:QTPTP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around T69 and T71 of human ATF2 (NP_001871.2?)
- Tested applications:IHC
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to ATF2 (phospho Thr69 + Thr71) for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC
- Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat