Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CFTR Rabbit Recombinant Antibody [clone: CFTR/7154R]
  ANTIA277961-100
New Product
undefined
Anti-CFTR Rabbit Recombinant Antibody [clone: CFTR/7154R]
  ANTIA277961-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ABC 35
  • Antigen symbol:
    CFTR
  • Clonality:
    Recombinant
  • Clone:
    CFTR/7154R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13569
  • Antigen synonyms:
    ABC35|ABCC7|ATP-binding cassette sub-family C member 7|cAMP-dependent chloride channel|cAMP dependent chloride channel|ATP binding cassette transporter sub family C member 7|ABCC 7|ATP binding cassette sub family C member 7|ATP Binding Cassette Superfamily C Member 7
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    165 - 170 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 258-385, of human CFTR protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [CFTR/7154R] antibody to CFTR for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CFTR
Clonality: Recombinant
Clone: CFTR/7154R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human