Anti-Myogenin Mouse Recombinant Antibody [clone: rMYOG/6297]
Catalog # ANTIA277905-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:bHLHc3
- Antigen symbol:Myogenin
- Clonality:Recombinant
- Clone:rMYOG/6297
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P15173
- Antigen synonyms:Class C basic helix-loop-helix protein 3|Myogenic factor 4|MYF 4|MYF4|Myogenin myogenin factor 4|MYOG|cb553|Myogenic factor 4 myogenin|Myf-4
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:34 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Myogenin protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rMYOG/6297] antibody to Myogenin for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Myogenin
Clonality: Recombinant
Clone: rMYOG/6297
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human