Anti-IRF3 Mouse Monoclonal Antibody [clone: PCRP-IRF3-3B2]
ANTIA277870-100
New Product
- Antibody type:Primary
- Antigen name:IIAE7
- Antigen symbol:IRF3
- Clonality:Monoclonal
- Clone:PCRP-IRF3-3B2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q14653
- Antigen synonyms:IRF3_HUMAN|Interferon regulatory factor 3|IRF3|MGC94729|IRF-3|IRF 3
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human IRF3 protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-IRF3-3B2] antibody to IRF3 for WB and IF with samples derived from human.
- Validated Applications: WB, IF
- Recommended Dilutions: WB: 2 to 4 µg/ml, IF: 1 to 2 µg/ml
Type: Primary
Antigen: IRF3
Clonality: Monoclonal
Clone: PCRP-IRF3-3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human