Anti-BrdU Rabbit Recombinant Antibody [clone: BRD/1539R]
ANTIA254205-100
New Product
- Antibody type:Primary
- Antigen name:BrdU
- Antigen symbol:BrdU
- Clonality:Recombinant
- Clone:BRD/1539R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:Liquid
- Antigen synonyms:BUdr|Bromodeoxyuridine
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:307Da
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bromodeoxyuridine conjugated to KLH.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [BRD/1539R] antibody to BrdU for ELISA.
- Validated Applications: ELISA
- Host: Rabbit
- Clonality: Monoclonal
- Iso
Type: Primary
Antigen: BrdU
Clonality: Recombinant
Clone: BRD/1539R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: