Anti-ds DNA Rabbit Recombinant Antibody [clone: DSD/4054R]
Catalog # ANTIA254201-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:ds DNA
- Antigen symbol:ds DNA
- Clonality:Recombinant
- Clone:DSD/4054R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Nuclei of Burkitt's cells.
- Tested applications:IHC-P
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [DSD/4054R] antibody to ds DNA for Flow Cytometry, IF and IHC-P with samples derived from human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ds DNA
Clonality: Recombinant
Clone: DSD/4054R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human