Anti-BrdU Mouse Recombinant Antibody [clone: rBRD494]
Catalog # ANTIA254040-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BrdU
- Antigen symbol:BrdU
- Clonality:Recombinant
- Clone:rBRD494
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Form:Liquid
- Antigen synonyms:BUdr|Bromodeoxyuridine
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bromodeoxyuridine conjugated to KLH.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rBRD494] antibody to BrdU for IHC-P.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
- Host: Mouse
- Clonality: Monoclonal
- Iso
Type: Primary
Antigen: BrdU
Clonality: Recombinant
Clone: rBRD494
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: