Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CD137 Rabbit Recombinant Antibody [clone: DM66]

Specifications

  • Catalog No:
  • ANTIA317400-500
  • ANTIA317400-100
  • Antigen symbol:
  • CD137
  • CD137
  • Antigen name:
  • CD137
  • CD137
  • Antigen synonyms:
  • CD137 antigen|CD137|CD 137|4 1BB|CDw137|4 1BB ligand receptor|4-1BB Ligand Receptor T Cell|4-1BB, mouse, homolog of|4-1BB ligand receptor|Antigen 4-1BB Homolog
  • CD137 antigen|CD137|CD 137|4 1BB|CDw137|4 1BB ligand receptor|4-1BB Ligand Receptor T Cell|4-1BB, mouse, homolog of|4-1BB ligand receptor|Antigen 4-1BB Homolog
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DM66
  • DM66
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • UniprotID# Q07011
  • UniprotID# Q07011
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Storage temperature:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together