Anti-PDGFB Mouse Monoclonal Antibody [Clone: PDGFB/3072]
ANTIA278327-100
New Product
- Antibody type:Primary
- Antigen symbol:PDGFB
- Clonality:Monoclonal
- Clone:PDGFB/3072
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2c kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01127
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:14 kDa (monomeric B chain)/31 - 35 kDa (dimer)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 27-158, of human PDGFB protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PDGFB/3072] antibody to PDGFB for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: PDGFB
Clonality: Monoclonal
Clone: PDGFB/3072
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human