Anti-Testosterone Mouse Monoclonal Antibody [Clone: 4E1G2]
Catalog # ANTIA254042-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:17b hydroxy 4 androsten 3 one
- Antigen symbol:Testosterone
- Clonality:Monoclonal
- Clone:4E1G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Form:Liquid
- Antigen synonyms:Testosterone|17ß-hydroxyandrost-4-ene-3-one
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:288,42Da
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Testosterone 3 CMO conjugated to BSA.
- Tested applications:Radioimmunoassay, IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [4E1G2] antibody to Testosterone for ELISA, Radioimmunoassay and IHC-P.
- Validated Applications: ELISA, Radioimmunoassay, IHC-P
- Recommended Dilutions: IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: Testosterone
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: