Anti-CD28 Mouse Monoclonal Antibody [Clone: C28/76]
Catalog # ANTIA253815-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 28
- Antigen symbol:CD28
- Clonality:Monoclonal
- Clone:C28/76
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P10747
- Antigen synonyms:CD28|T cell specific surface glycoprotein CD28|MGC138290|T-cell-specific surface glycoprotein CD28|CD28_HUMAN|T cell specific surface glycoprotein|CD28 molecule|CD28 antigen|T cell antigen CD28
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:44 kDa (monomer)/88 kDa (dimer)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human CD28 protein.
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C28/76] antibody to CD28 for Flow Cytometry and Functional Studies with samples derived from human.
- Validated Applications: Flow Cytometry, Functional Studies
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: C28/76
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human