Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-RSK1 p90 (phospho Thr348) Rabbit Polyclonal Antibody
  ANTIA309559-100
New Product
undefined
Anti-RSK1 p90 (phospho Thr348) Rabbit Polyclonal Antibody
  ANTIA309559-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    90 kDa ribosomal protein S6 kinase 1
  • Antigen symbol:
    RSK1 p90
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q15418
  • Antigen synonyms:
    MAPK-activated protein kinase 1a|MAP kinase-activated protein kinase 1a|EC 2.7.11.1|dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1|HU 1|MAP kinase activated protein kinase 1a|dJ590P13.1|HU1|KS6A1_HUMAN
  • Modification:
    phospho Thr348
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
  • Molecular weight:
    83 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around T348 of human p90Rsk/RSK1/RPS6KA1.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to RSK1 p90 (phospho Thr348) for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: RSK1 p90
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat