Anti-Serine racemase Rabbit Monoclonal Antibody [clone: ARC3047]
Catalog # ANTIA309468-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:D serine ammonia lyase
- Antigen symbol:Serine racemase
- Clonality:Monoclonal
- Clone:ARC3047
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9GZT4
- Antigen synonyms:L serine ammonia lyase|L-serine dehydratase|D serine dehydratase|ISO1|ILV1|L-serine ammonia-lyase|D-serine dehydratase|D-serine ammonia-lyase|L serine dehydratase
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of Human Serine racemase.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC3047] antibody to Serine racemase for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Serine racemase
Clonality: Monoclonal
Clone: ARC3047
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human