Order Entry
United Kingdom
ContactUsLinkComponent
Anti-alpha Internexin Mouse Monoclonal Antibody [clone: 2E3]
Anti-alpha Internexin Mouse Monoclonal Antibody [clone: 2E3]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-alpha Internexin Mouse Monoclonal Antibody [clone: 2E3]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA85448-500
  • ANTIA85448-250
  • Antigen symbol:
  • alpha Internexin
  • alpha Internexin
  • Antigen name:
  • 66 kDa neurofilament protein
  • 66 kDa neurofilament protein
  • Antigen synonyms:
  • Alpha Inx|Alpha-internexin|MGC12702|INA|Alpha-Inx|alpha Internexin|AINX_HUMAN|Internexin neuronal intermediate filament protein alpha|NEF 5
  • Alpha Inx|Alpha-internexin|MGC12702|INA|Alpha-Inx|alpha Internexin|AINX_HUMAN|Internexin neuronal intermediate filament protein alpha|NEF 5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2E3
  • 2E3
  • Reactivity:
  • Human, Bovine, Rat, Mouse
  • Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# Q16352
  • UniprotID# Q16352
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant full-length rat alpha Internexin, expressed in and purified from E. coli
  • Recombinant full-length rat alpha Internexin, expressed in and purified from E. coli
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 64 - 66 kDa (by SDS-PAGE)
  • 64 - 66 kDa (by SDS-PAGE)
  • Purification:
  • Immunogen affinity purification.
  • Immunogen affinity purification.
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
  • Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
  • Storage temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at +4 °C
  • Shipped on blue ice at +4 °C
  • Tested applications:
  • ICC/IF, IHC
  • ICC/IF, IHC

Specifications

Frequently Bought Together