Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Adrenomedullin Rabbit Polyclonal Antibody
  ANTIA98324-100
New Product
undefined
Anti-Adrenomedullin Rabbit Polyclonal Antibody
  ANTIA98324-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Adm
  • Antigen symbol:
    Adrenomedullin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P35318
  • Antigen synonyms:
    ADM precursor|Proadrenomedullin N20 terminal peptide|AM|Proadrenomedullin N-20 terminal peptide|ADML_HUMAN|Adrenomedullin|PAMP|ProAM N terminal 20 peptide|Preproadrenomedullin
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    20 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human ADM (amino acids 51-100)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to ADM for IHC and ELISA with samples derived from human, mouse and rat.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: Adrenomedullin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat