Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ATPG Rabbit Polyclonal Antibody
Anti-ATPG Rabbit Polyclonal Antibody
Catalog # ANTIA98278-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-ATPG Rabbit Polyclonal Antibody
Catalog # ANTIA98278-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP synthase gamma chain, mitochondrial
  • Antigen symbol:
    ATPG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P36542
  • Antigen synonyms:
    ATP5C1|ATP5C 1|ATP5C|ATP5CL1|ATP synthase subunit gamma, mitochondrial|ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1|ATPG|ATP synthase, H+ transporting, mitochondrial F1 complex, gamma subunit|ATP synthase subunit gamma
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    32 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human ATP5C1 (amino acids 131-180)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to ATPG for WB and ELISA with samples derived from human, mouse and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000

Type: Primary
Antigen: ATPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat