Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-BRAF35 Rabbit Polyclonal Antibody
  ANTIA307782-100
New Product
undefined
Anti-BRAF35 Rabbit Polyclonal Antibody
  ANTIA307782-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    BRAF25
  • Antigen symbol:
    BRAF35
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9P0W2
  • Antigen synonyms:
    HMG domain-containing protein HMGX2|BRCA2 associated factor 35|High Mobility Group Protein 20B|BRCA2-associated factor 35|HMG box containing protein 20B|HM20B_HUMAN|BRAF35|HMG box-containing protein 20B|HMG domain-containing protein 2
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Molecular weight:
    36 kDa
  • Sequence:
    MSHGPKQPGAAAAPAGGKAPGQHGGFVVTVKQERGEGPRAGEKGSHEEEPVKKRGWPKGKKRKKILPNGPKAPVTGYVRFLNERREQIRTRHPDLPFPEITKMLGAEWSKLQPTEKQRYLDEAEREKQQYMKELRAYQQSEAYKMCTEKIQEKKIKKEDSSSGLMNTLLNGHKGGDCDGFSTFDVPIFTEEFLDQNKAREAELRRLRKMNVAFEEQNAVLQRHTQSMSSARERLEQELALEERRTLALQQQLQAVRQALTASFASLPVPGTGETPTLGTLDFYMARLHGAIERDPAQHEKLIVRIKEILAQVASEHL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human HMG20B (NP_006330.2)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to BRAF35 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:200 to 1:2000

Type: Primary
Antigen: BRAF35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat