Specifications
- Catalog No:
- ANTIA318570-500
- ANTIA318570-100
- Antigen symbol:
- 5T4
- 5T4
- Antigen synonyms:
- AW495680|5T4 oncotrophoblast glycoprotein|TPBG_HUMAN|M6P1|5T4AG|TPBG|5T4 oncofetal trophoblast glycoprotein|5T4 oncofetal antigen
- AW495680|5T4 oncotrophoblast glycoprotein|TPBG_HUMAN|M6P1|5T4AG|TPBG|5T4 oncofetal trophoblast glycoprotein|5T4 oncofetal antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM137
- DM137
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# Q13641
- UniprotID# Q13641
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together