Anti-SLC6A12 Goat Polyclonal Antibody
Catalog # ANTIA83618-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Betaine/GABA transporter 1
- Antigen symbol:SLC6A12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:BGT1|S6A12_HUMAN|Sodium- and chloride-dependent betaine transporter|Solute carrier family 6 (neurotransmitter transporter), member 12|BGT-1|Na(+)/Cl(-) betaine/GABA transporter|GAT2|SLC6A12|Gamma aminobutyric acid transporter
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:CLDGSAGRNFGPSPT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human SLC6A12 (C terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to SLC6A12 for ELISA, WB and IHC with samples derived from human.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: SLC6A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human