Anti-RBP4 Mouse Monoclonal Antibody [clone: RBP4/4053]
Catalog # ANTIA253023-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:OTTHUMP00000020114
- Antigen symbol:RBP4
- Clonality:Monoclonal
- Clone:RBP4/4053
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2c kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P02753
- Antigen synonyms:RBP4|Plasma retinol-binding protein|OTTHUMP00000020115|RDCCAS|Plasma retinol binding protein 4|Plasma retinol-binding protein(1-176)|OTTHUMP00000020116|PRO2222|PRBP
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:25 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 29 - 148, of human RBP4 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [RBP4/4053] antibody to RBP4 for IHC-P with samples derived from Human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: RBP4/4053
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human