Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-PCNA Mouse Monoclonal Antibody [clone: SPM350]
  ANTIA252759-100
New Product
undefined
Anti-PCNA Mouse Monoclonal Antibody [clone: SPM350]
  ANTIA252759-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ATLD2
  • Antigen symbol:
    PCNA
  • Clonality:
    Monoclonal
  • Clone:
    SPM350
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Zebrafish,
    S. cerevisiae,
    Pig,
    Drosophila melanogaster,
    Monkey,
    S. pombe,
    Chicken,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P12004
  • Antigen synonyms:
    DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    36 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Rat PCNA/protein A fusion protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [SPM350] antibody to PCNA for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Monkey, Porcine, Mouse, Rat, Chicken, Zebrafish, Drosophila melanogaster, S. pombe and S. cerevisiae.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: SPM350
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Monkey;Porcine;Mouse;Rat;Chicken;Zebrafish;Drosophila melanogaster;S. pombe;S. cerevisiae