Anti-Galectin 1 Mouse Monoclonal Antibody [clone: GAL1/1831]
Catalog # ANTIA252406-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:14 kDa laminin-binding protein
- Antigen symbol:Galectin 1
- Clonality:Monoclonal
- Clone:GAL1/1831
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2 lambda
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P09382
- Antigen synonyms:Beta galactoside binding lectin|GAL1|beta galactoside binding protein|14 kDa lectin|Beta-galactoside-binding lectin L-14-I|Beta galactoside binding lectin L 14 I|Gal-1|Gal 1|DKFZp686E23103
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:14 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 12 - 108, of human Galectin 1 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [GAL1/1831] antibody to Galectin 1 for WB, ELISA, IF and IHC-P with samples derived from Human.
- Validated applications: WB, ELISA, IF, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Galectin 1
Clonality: Monoclonal
Clone: GAL1/1831
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 lambda
Reactivity: Human