Anti-Lambda Light Chain Mouse Recombinant Antibody [clone: rLLC/1738]
Catalog # ANTIA252179-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bence Jones protein
- Antigen symbol:Lambda Light Chain
- Clonality:Recombinant
- Clone:rLLC/1738
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01701, P01842
- Antigen synonyms:Ig lambda chain C regions|BJP|IGLC 3|ig lambda-6 chain C region|IGLC 1|IGLC|IGLC 2|IGLC1|Constant region of lambda light chains
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:~22,5 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified human lambda light chain
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rLLC/1738] antibody to Lambda Light Chain for IHC-P with samples derived from Human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Lambda Light Chain
Clonality: Recombinant
Clone: rLLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human