Anti-HSP60 Mouse Monoclonal Antibody [clone: HSPD1/780]
Catalog # ANTIA252067-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:60 kDa chaperonin
- Antigen symbol:HSP60
- Clonality:Monoclonal
- Clone:HSPD1/780
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Pig,Rabbit,Monkey,Sheep,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P10809
- Antigen synonyms:Chaperonin, 60-KD|GROEL|Heat shock 60kDa protein 1|Chaperonin 60|CH60_HUMAN|fa04a05|CPN60|60 kDa heat shock protein, mitochondrial|heat shock 60kDa protein 1 (chaperonin)
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:60 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human HSPD1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HSPD1/780] antibody to HSP60 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Mouse, Rat, Hamster, Sheep, Rabbit, Bovine, Canine, Porcine, Monkey and Chicken.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HSP60
Clonality: Monoclonal
Clone: HSPD1/780
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Hamster;Sheep;Rabbit;Bovine;Dog;Porcine;Monkey;Chicken