Order Entry
United Kingdom
ContactUsLinkComponent
Anti-EIF4A2 Mouse Monoclonal Antibody [clone: PCRP-EIF4A2-2B5]
Anti-EIF4A2 Mouse Monoclonal Antibody [clone: PCRP-EIF4A2-2B5]
Catalog # ANTIA251597-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-EIF4A2 Mouse Monoclonal Antibody [clone: PCRP-EIF4A2-2B5]
Catalog # ANTIA251597-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP dependent RNA helicase eIF4A 2
  • Antigen symbol:
    EIF4A2
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-EIF4A2-2B5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q14240
  • Antigen synonyms:
    eIF4A-II|eIF 4A II|EIF4A|eIF4A2|eIF4A II|BM-010|ATP-dependent RNA helicase eIF4A-2|eIF-4A-II|DDX2B
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human EIF4A2 protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [PCRP-EIF4A2-2B5] antibody to EIF4A2 for ELISA with samples derived from Human.

  • Validated applications: ELISA

Type: Primary
Antigen: EIF4A2
Clonality: Monoclonal
Clone: PCRP-EIF4A2-2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human