Anti-Carboxypeptidase A1 Mouse Monoclonal Antibody [clone: CPA1/2714]
Catalog # ANTIA251436-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Carboxypeptidase A
- Antigen symbol:Carboxypeptidase A1
- Clonality:Monoclonal
- Clone:CPA1/2714
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P15085
- Antigen synonyms:Procarboxypeptidase A1 pancreatic|carboxypeptidase A1 (pancreatic)|CPA1|Pancreatic carboxypeptidase A1|Carboxypeptidase A1|Carboxypeptidase A1 precursor|CBPA1_HUMANCPA
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:47 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CPA1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CPA1/2714] antibody to Carboxypeptidase A1 for WB and IHC-P with samples derived from Human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Carboxypeptidase A1
Clonality: Monoclonal
Clone: CPA1/2714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human