Anti-BrdU Rabbit Recombinant Antibody [clone: BRDU/3902R]
ANTIA251007-100
New Product
- Antibody type:Primary
- Antigen name:BrdU
- Antigen symbol:BrdU
- Clonality:Recombinant
- Clone:BRDU/3902R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Form:Liquid
- Antigen synonyms:BUdr|Bromodeoxyuridine
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bromodeoxyuridine conjugated to KLH
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [BRDU/3902R] antibody to BrdU for IHC-P.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
- Host: Rabbit
- Clonality: Monoclonal
- Isotype: IgG
- Conjugate: Unconjugated
Type: Primary
Antigen: BrdU
Clonality: Recombinant
Clone: BRDU/3902R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: