Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-BrdU Rabbit Recombinant Antibody [clone: BRD/1539R]
  ANTIA251026-100
New Product
undefined
Anti-BrdU Rabbit Recombinant Antibody [clone: BRD/1539R]
  ANTIA251026-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    BrdU
  • Antigen symbol:
    BrdU
  • Clonality:
    Recombinant
  • Clone:
    BRD/1539R
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Form:
    Liquid
  • Antigen synonyms:
    BUdr|Bromodeoxyuridine
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    307Da
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Bromodeoxyuridine conjugated to KLH
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [BRD/1539R] antibody to BrdU for ELISA.

  • Validated applications: ELISA
  • Host: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Conjugate: Unconjugated

Type: Primary
Antigen: BrdU
Clonality: Recombinant
Clone: BRD/1539R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: