Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 + BRD494 + BRD.3]
Catalog # ANTIA250997-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BrdU
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:BRD469 + BRD494 + BRD.3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:Multiple
- Form:Liquid
- Antigen synonyms:BUdr|Bromodeoxyuridine
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bromodeoxyuridine conjugated to BSA or KLH
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [BRD469 + BRD494 + BRD.3] antibody to BrdU for IHC-P.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
- Host: Mouse
- Clonality: Oligoclonal
- Isotype: IgG1, kappa + IgG1, kappa + IgG1, kappa
- Conjugate: Unconjugated
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: BRD469 + BRD494 + BRD.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: