Order Entry
United Kingdom
ContactUsLinkComponent
 

 

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid polyneuropathy
  • Antigen symbol:
    Transthyretin
  • Clonality:
    Monoclonal
  • Clone:
    TTR/4295
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02766
  • Antigen synonyms:
    Amyloidosis I|HEL111|CTS1|Dystransthyretinemic hyperthyroxinemia|Dysprealbuminemic euthyroidal hyperthyroxinemia|CTS|Carpal tunnel syndrome 1|ATTR|Epididymis luminal protein 111
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    15 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human Transthyretin protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [TTR/4295] antibody to Transthyretin for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 2 to 4 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Transthyretin
Clonality: Monoclonal
Clone: TTR/4295
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human