Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Transcription factor AP-2-alpha Rabbit Polyclonal Antibody
Anti-Transcription factor AP-2-alpha Rabbit Polyclonal Antibody
Catalog # ANTIA11149-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Transcription factor AP-2-alpha Rabbit Polyclonal Antibody
Catalog # ANTIA11149-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Transcription factor AP-2-alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05549
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    50 kDa
  • Sequence:
    GLDPRRDYRRHEDLLHGPHALSSGLGDLSIHSLPHAIEEVPHVEDPGINIPDQTVIKKGPVSLSKSNSNAVSAIPINKDNL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 120 - 200 of human Activator protein 2 (AP - 2/TFAP2A) (NP_003211.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Transcription factor AP-2-alpha for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: Transcription factor AP-2-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat