Order Entry
United Kingdom
ContactUsLinkComponent
Anti-LI Cadherin Rabbit Recombinant Antibody [clone: CDH17/8158R]
Anti-LI Cadherin Rabbit Recombinant Antibody [clone: CDH17/8158R]
Catalog # ANTIA316633-100
Supplier:  ANTIBODIES.COM
New Product
Anti-LI Cadherin Rabbit Recombinant Antibody [clone: CDH17/8158R]
Catalog # ANTIA316633-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BILL-cadherin
  • Antigen symbol:
    LI Cadherin
  • Clonality:
    Recombinant
  • Clone:
    CDH17/8158R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q12864
  • Antigen synonyms:
    CDH16|CAD17_HUMAN|Cadherin-17|cadherin, liver-intestine|cadherin 17, LI cadherin (liver-intestine)|Cadherin 16, formerly|CDH16, formerly|Cadherin 16|Cadherin 17
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    120 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 242-418) of human Cadherin 17 protein (CDH17). The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [CDH17/8158R]] antibody to LI Cadherin for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: LI Cadherin
Clonality: Recombinant
Clone: CDH17/8158R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human