Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ds DNA Rabbit Recombinant Antibody [clone: DSD/8204R]
Anti-ds DNA Rabbit Recombinant Antibody [clone: DSD/8204R]
Catalog # ANTIA316281-100
Supplier:  ANTIBODIES.COM
New Product
Anti-ds DNA Rabbit Recombinant Antibody [clone: DSD/8204R]
Catalog # ANTIA316281-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ds DNA
  • Clonality:
    Recombinant
  • Clone:
    DSD/8204R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Nuclei of Burkitt's cells.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [DSD/8204R] antibody to ds DNA for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: ds DNA
Clonality: Recombinant
Clone: DSD/8204R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human