Anti-Aciculin Mouse Recombinant Antibody [clone: rPGM5/7164]
Catalog # ANTIA315816-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Aciculin
- Clonality:Recombinant
- Clone:rPGM5/7164
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q15124
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:56 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human PGM5 protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rPGM5/7164] antibody to Aciculin for ELISA, IF and WB with samples derived from human.
- Validated applications: ELISA, IF, WB
- Recommended dilutions: IF: 1 to 2 µg/ ml, WB: 1 to 2 µg/ ml, IHC: 2 to 4 µg/ ml
Type: Primary
Antigen: Aciculin
Clonality: Recombinant
Clone: rPGM5/7164
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human