Anti-PD1 Mouse Monoclonal Antibody [clone: PDCD1/7125]
Catalog # ANTIA315488-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD279
- Antigen symbol:PD1
- Clonality:Monoclonal
- Clone:PDCD1/7125
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q15116
- Antigen synonyms:hPD 1|hPD-1|PDCD1_HUMAN|PDCD 1|hPD l|hSLE1|PDCD1|PD1|CD279 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:55 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human PDCD1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PDCD1/7125] antibody to PD1 for ELISA and IHC-P with samples derived from human.
- Validated applications: ELISA, IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: PDCD1/7125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human