Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Ki67 Mouse Monoclonal Antibody [clone: MKI67/8743]
Anti-Ki67 Mouse Monoclonal Antibody [clone: MKI67/8743]
Catalog # ANTIA315219-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Ki67 Mouse Monoclonal Antibody [clone: MKI67/8743]
Catalog # ANTIA315219-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    Ki67
  • Clonality:
    Monoclonal
  • Clone:
    MKI67/8743
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P46013
  • Antigen synonyms:
    MIB 1|MIB|Antigen KI-67|MKI67|KIA|Marker of proliferation Ki-67|KI67_HUMAN|Antigen KI67|PPP1R105
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    345 kDa/395 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human Ki67 protein fragment (around Amino Acids 1-200). The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MKI67/8743] antibody to Ki67 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Ki67
Clonality: Monoclonal
Clone: MKI67/8743
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human