Anti-TL1A Recombinant Recombinant Antibody [clone: DMC464]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Recombinant chimeric monoclonal [DMC464] antibody to TL1A for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: TL1A
Clonality: Recombinant
Clone: DMC464
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ANTIA318726-500
- ANTIA318726-100
- Antigen symbol:
- TL1A
- TL1A
- Antigen name:
- MGC129934
- MGC129934
- Antigen synonyms:
- TNF ligand-related molecule 1|TL1A|tl1|tnf superfamily ligand tl1a|TNFSF15|TL1 A|tnf ligand related molecule 1|TNF15_HUMAN|MGC129935
- TNF ligand-related molecule 1|TL1A|tl1|tnf superfamily ligand tl1a|TNFSF15|TL1 A|tnf ligand related molecule 1|TNF15_HUMAN|MGC129935
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC464
- DMC464
- Reactivity:
- Human
- Human
- Host:
- Recombinant
- Recombinant
- Gene ID:
- UniprotID# O95150
- UniprotID# O95150
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Storage temperature:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together